Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions EDTA Titrant, 0.01 Molar (0.02 Normal) (4L)
SDP

Catalog No. NC9434564
Encompass_Preferred

EDTA Titrant, 0.01 Molar (0.02 Normal) (4L)

Catalog No. NC9434564 Supplier Aqua Solutions Supplier No. 35504L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EDTA Titrant, 0.01 Molar (0.02 Normal) (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.