Learn More
EDTA Titrant, 0.0200 Molar (M/50), Reagents
Specifications
Specifications
| Chemical Name or Material | EDTA Titrant, 0.0200 Molar (M/50) |
| Color | Colorless |
| CAS | 7732-18-5, 6381-92-6 |
| Boiling Point | 100°C |
| Molecular Formula | C10H14N2O8Na2/H2O |
| Synonym | Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.003 |
| pH | 4 to 6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.