Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.0200 Molar (M/50), Reagents

EDTA Titrant, 0.0200 Molar (M/50), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$39.75 - $61.50

Specifications

Chemical Name or Material EDTA Titrant, 0.0200 Molar (M/50)
Color Colorless
CAS 7732-18-5 , 6381-92-6
Boiling Point 100°C
Molecular Formula C10H14N2O8Na2/H2O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-335-380
View Documents
Reagents Holdings Llc
CE1050001A
1 L
Each for $61.50
Add to Cart
 
 Restricted Item
01-335-378
View Documents
Reagents Holdings Llc
CE105000500A
500 mL
Each for $39.75
Add to Cart
 
 Restricted Item
Description

Description

Specifications

Specifications

EDTA Titrant, 0.0200 Molar (M/50)
7732-18-5 , 6381-92-6
C10H14N2O8Na2/H2O
Reagent
4 to 6
Liquid
Colorless
100°C
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
1.003
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.