Learn More
EDTA Titrant, 0.0500 Molar (M/20), Reagents
EDTA Titrant, 0.0500 Molar (M/20), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier:  Reagents Holdings Llc CE2530001A
 View more versions of this product
Catalog No. 01-335-331
 Restricted Item
Add to Cart
Description
Description
Specifications
Specifications
EDTA Titrant, 0.0500 Molar (M/20)
7732-18-5 , 6381-92-6
C10H14N2O8Na2/H2O
Reagent
1.009
1 L
Liquid
Colorless
100°C
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
Poly Bottle
4 to 6
Non-sterile
Product Suggestions
Product Suggestions