Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.05M solution, Macron Fine Chemicals™

Catalog No. MKH1154 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-H115-4 4 L
MK-H115-1 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-H115-4 Supplier Avantor Supplier No. H11507
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
Density 860mg/mL
Formula Weight 292.24
Quantity 4 L
Physical Form Crystals
Chemical Name or Material EDTA Titrant, 0.01M solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.