Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.100 M with Magnesium, Reagents

Catalog No. 01335361
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-361 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-361 Supplier Reagents Holdings Llc Supplier No. CE2502001A
Add to Cart
Add to Cart

EDTA Titrant, 0.100 M with Magnesium, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material EDTA Titrant, 0.100 M with Magnesium
Color Colorless
CAS 7732-18-5, 6381-92-6
Boiling Point 100°C
Molecular Formula C10H14N2O8Na2/H2O
Synonym Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.018
pH 4 to 6
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.