Learn More
EDTA Titrant, 0.100 M with Magnesium, Reagents
EDTA Titrant, 0.100 M with Magnesium, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$55.80
Specifications
| Chemical Name or Material | EDTA Titrant, 0.100 M with Magnesium |
|---|---|
| Color | Colorless |
| CAS | 7732-18-5 , 6381-92-6 |
| Boiling Point | 100°C |
| Molecular Formula | C10H14N2O8Na2/H2O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-361
|
Reagents Holdings Llc
CE2502001A |
1 L |
Each of 1 for $55.80
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.