Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediaminetetraacetic acid trisodium salt hydrate, 85715-60-2, MFCD00149675, 1 kg
Catalog No. NC1259019
Encompass
Empirical Formula (Hill Notation): C10H13N2Na3O8 · xH2O, Molecular Weight: 358.19 (anhydrous basis), ≥95%, Synonym: (Ethylenedinitrilo)tetraacetic acid trisodium salt hydrate, EDTA trisodium salt hydrate, Edathamil hydrate. Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.
Catalog No. NC1259019 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ED3SS1KG
Empirical Formula (Hill Notation): C10H13N2Na3O8 · xH2O, Molecular Weight: 358.19 (anhydrous basis), ≥95%, Synonym: (Ethylenedinitrilo)tetraacetic acid trisodium salt hydrate, EDTA trisodium salt hydrate, Edathamil hydrate. Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.