Learn More
Apexbio Technology LLC EdU Cell Proliferation Kit (TMB) 500tests
SDP
Supplier:  Apexbio Technology LLC K22792000
Encompass_Preferred
EdU Flow Cytometry Assay Kits (HF594) provides a novel method based on click chemistry specifically CuAAC (copper-catalyzed azide-alkyne cycloaddition) enabling direct measurement of DNA synthesis during the S-phase of the cell cycle The thymidine analog EdU (5-ethynyl-2 -deoxyuridine) can be incorporated directly into DNA strands during DNA synthesis The alkyne group on EdU a biologically inert moiety undergoes highly selective reaction with azide-functionalized dyes via the CuAAC reaction forming a stable 1 2 3-triazole product The EdU Cell Proliferation Kit (TMB) specifically targets proliferating cell DNA by linking EdU with biotin azide followed by incubating with HRP-Streptavidin which binds to biotin The bound HRP then catalyzes a colorimetric reaction with TMB substrate allowing final quantification by spectrophotometric detection This kit is designed for use with cell samples and is not suitable for tissue section analysis
Catalog No. 50-310-7070