Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Edu-Click 647

Catalog No. BCKEDU647 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Catalog No. BCKEDU647 Supplier Sigma Aldrich Supplier No. BCKEDU647
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog to thymidine and is incorporated into DNA during active DNA synthesis. In contrast to BrdU assays, the EdU-Click Assays are not antibody based and therefore do not require DNA denaturation for detection of the incorporated nucleoside. Instead, the Baseclick EdU-Click Assays utilize click chemistry for detection in a variety of dye fluorescent readouts. Furthermore, the streamlined detection protocol reduces both the total number of steps and significantly decreases the total amount of time. The simple click chemistry detection procedure is complete within 30 minutes and is compatible with multiplexing for content and context-rich results.Imaging

Specifications

CAS 89-77-0
Synonym Baran Olefin Cross-Coupling Catalyst; Iron tris(diisobutyrylmethane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.