Learn More
Apexbio Technology LLC EdU Flow Cytometry Assay Kits (HF488) 100tests
SDP
Catalog No. 503107303
Encompass_Preferred
EdU Flow Cytometry Assay Kits (HF488) provides a novel method based on click chemistry specifically CuAAC (copper-catalyzed azide-alkyne cycloaddition) enabling direct measurement of DNA synthesis during the S-phase of the cell cycle The thymidine analog EdU (5-ethynyl-2 -deoxyuridine) can be incorporated directly into DNA strands during DNA synthesis The alkyne group on EdU a biologically inert moiety undergoes highly selective reaction with azide-functionalized dyes via the CuAAC reaction forming a stable 1 2 3-triazole product The EdU Flow Cytometry Assay Kits (HF488) use HyperFluor 488 azide to label the DNA of proliferating cells following its reaction with EdU Samples labeled in this manner can be analyzed via flow cytometry (HyperFluor 594 azide excitation maximum at 496 nm emission maximum at 516 nm)
Catalog No. 50-310-7303 Supplier Apexbio Technology LLC Supplier No. K2241100
Add to Cart
Add to Cart
EdU Flow Cytometry Assay Kits (HF488) provides a novel method based on click chemistry specifically CuAAC (copper-catalyzed azide-alkyne cycloaddition) enabling direct measurement of DNA synthesis during the S-phase of the cell cycle The thymidine analog EdU (5-ethynyl-2 -deoxyuridine) can be incorporated directly into DNA strands during DNA synthesis The alkyne group on EdU a biologically inert moiety undergoes highly selective reaction with azide-functionalized dyes via the CuAAC reaction forming a stable 1 2 3-triazole product The EdU Flow Cytometry Assay Kits (HF488) use HyperFluor 488 azide to label the DNA of proliferating cells following its reaction with EdU Samples labeled in this manner can be analyzed via flow cytometry (HyperFluor 594 azide excitation maximum at 496 nm emission maximum at 516 nm)