Learn More
Apexbio Technology LLC EdU Flow Cytometry Assay Kits (HF594) 100tests
SDP
Catalog No. 503107145
Encompass_Preferred
EdU Flow Cytometry Assay Kits (HF594) provides a novel method based on click chemistry specifically CuAAC (copper-catalyzed azide-alkyne cycloaddition) enabling direct measurement of DNA synthesis during the S-phase of the cell cycle The thymidine analog EdU (5-ethynyl-2 -deoxyuridine) can be incorporated directly into DNA strands during DNA synthesis The alkyne group on EdU a biologically inert moiety undergoes highly selective reaction with azide-functionalized dyes via the CuAAC reaction forming a stable 1 2 3-triazole product The EdU Flow Cytometry Assay Kits (HF594) use HyperFluor 594 azide to label the DNA of proliferating cells following its reaction with EdU Samples labeled in this manner can be analyzed via flow cytometry (HyperFluor 594 azide excitation maximum at 590 nm emission maximum at 617 nm)
Catalog No. 50-310-7145 Supplier Apexbio Technology LLC Supplier No. K2244100
EdU Flow Cytometry Assay Kits (HF594) provides a novel method based on click chemistry specifically CuAAC (copper-catalyzed azide-alkyne cycloaddition) enabling direct measurement of DNA synthesis during the S-phase of the cell cycle The thymidine analog EdU (5-ethynyl-2 -deoxyuridine) can be incorporated directly into DNA strands during DNA synthesis The alkyne group on EdU a biologically inert moiety undergoes highly selective reaction with azide-functionalized dyes via the CuAAC reaction forming a stable 1 2 3-triazole product The EdU Flow Cytometry Assay Kits (HF594) use HyperFluor 594 azide to label the DNA of proliferating cells following its reaction with EdU Samples labeled in this manner can be analyzed via flow cytometry (HyperFluor 594 azide excitation maximum at 590 nm emission maximum at 617 nm)