Learn More
Apexbio Technology LLC EdU Imaging Kits (Cy5), 50-500 tests. EdU Imaging Kits (Cy5) is a sensitive and reliable way to detect and quantify cell proliferation in cells.
SDP
Catalog No. 501998210
Encompass_Preferred
EdU Imaging Kit (Cy5) is a sensitive and reliable way to detect and quantify cell proliferation in mammalian cells. It is optimized for fluorescence microscopy. It is a better alternative to traditional BrdU (bromodeoxyuridine). Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8210 Supplier Apexbio Technology LLC Supplier No. K107650
EdU Imaging Kit (Cy5) is a sensitive and reliable way to detect and quantify cell proliferation in mammalian cells. It is optimized for fluorescence microscopy. It is a better alternative to traditional BrdU (bromodeoxyuridine). Other sizes are also available. Please inqury us for quote.