Learn More
Azzota Corp Edu Vis Spectro WL 330-1000nm
Catalog No. 5000886994
Encompass_Preferred
Education Vis Spectrophotometer WL rng 330-1000nm Spectral slit width 6nm WL accuracy 2nm WL readability 1nm Stray radiant energy Less than 0 3%T at 340nm at 400nm Transmittance 0%to 125 0%T Absorbency 0A to 1 999A concentration 0-1999C(0-1999F) Photometric accuracy 1 0%T 110V 60Hz
Catalog No. 50-008-86994 Supplier Azzota Corp Supplier No. SV800
Education Vis Spectrophotometer WL rng 330-1000nm Spectral slit width 6nm WL accuracy 2nm WL readability 1nm Stray radiant energy Less than 0 3%T at 340nm at 400nm Transmittance 0%to 125 0%T Absorbency 0A to 1 999A concentration 0-1999C(0-1999F) Photometric accuracy 1 0%T 110V 60Hz