Learn More
Bachem H-3-Iodo-Tyr-OH
SDP
Supplier:  Bachem 4026261.0025
Encompass_Preferred
H-3-Iodo-Tyr-OH, 70-78-0, C9H10INO3, Mr 307.09, 25g. Educt (after protection with Z/Bzl) for the synthesis of dityrosine via Miyaura borylation followed by Suzuki coupling. The appraoch is also suitable for incorporating dityrosine crosslinks into peptides. Inhibitor of tyrosine hydroxylase.
Catalog No. 50-194-7082