Learn More
Bachem Fmoc-p-bromo-Phe-OH
SDP
Supplier:  Bachem 4042637.0005
Encompass_Preferred
Fmoc-p-bromo-Phe-OH, 198561-04-5, C24H20BrNO4, Mr 466.33, 5g. Educt (coupled to Wang resin) for obtaining a library of p-biphenylalanines via Suzuki coupling.
Catalog No. 50-194-7260