Learn More
Bachem Fmoc-Ser-OH · H2O
SDP
Supplier:  Bachem 4044985.0250
Encompass_Preferred
Fmoc-Ser-OH · H2O, 73724-45-5, C18H17NO5 · H2O, Mr 345.35, 250g. Educt for the synthesis of O-glycosylated serine building blocks. Liu et al. synthesized O-mannosylated Fmoc-Ser.
Catalog No. 50-194-7302