Learn More
Bachem Fmoc-Homocys(Trt)-OH
SDP
Supplier:  Bachem 4025118.0005
Encompass_Preferred
Fmoc-Homocys(Trt)-OH, 167015-23-8, C38H33NO4S, Mr 599.75, 5g. (S)-2-(Fmoc-amino)-4-tritylsulfanyl-butyric acid. Educt for the synthesis of b-turn peptidomimetics (Jiang and Burgess). Lindman et al. obtained Hcy-containing angiotensin II analogs by Fmoc-SPPS. Homocysteine can function as thiol component during chemical ligation. S-Methylation of the resulting peptide yields methionine.
Catalog No. 50-194-7202