Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ 3.0 L Digital Water Bath
SDPSCI_ED

Catalog No. S37220 Shop All EDVOTEK Products
Change view
Click to view available options
Capacity (Metric):
3.0 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S37220 3.0 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37220 Supplier Edvotek™ Supplier No. 539
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Cover

One-piece seamless, stainless-steel chamber is corrosion resistant.

  • Digital temperature control
  • Low-water sensor prevents burnouts
  • Deep chamber holds bottles and flasks
  • Ideal for enzyme incubations
  • Maximum temperature: 95°C (203°F)
  • Temperature accuracy: ±1°C
  • Chamber size: 16 x 16 x 13 cm
  • Built-in timer

Specifications

Controller Type Digital
Capacity (Metric) 3.0 L
Temperature Range 95°C
Type Water Bath
Includes Cover is included
Material Stainless Steel
Power Consumption 300W, 110V, 50 Hz
Temperature Accuracy ±1°C
Shipping Weight (Metric) 2.13 kg
Electrical Requirements 100 to 130 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.