Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Advanced Placement Laboratory Kit 12: Dissolved Oxygen and Aquatic Primary Productivity
SDPSCI_ED

Catalog No. S11292 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11292 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11292 Supplier Edvotek™ Supplier No. 292
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, sodium iodide, sodium hydroxide, manganese sulfate, starch solution, sodium thiosulfate, nitrogen enrichment solution, phosphorous enrichment reagents, transfer pipets, microcentrifuge tubes, and 15 ml conical tubes

Requires

  • Pond/lake water
  • Goggles
  • Grow lights
  • Pipets
  • Sulfuric acid (concentrated)
  • Plastic window screens
  • Burets
  • 50 Sealable containers or BOD bottles
  • Flasks
  • Beakers
  • Gloves
  • Filter paper
  • Linear graph paper
  • Aluminum foil

Experiment demonstrates the methodology used for measuring dissolved oxygen and determining the primary productivity of a natural body of water.

Dissolved oxygen levels are used to monitor the health and productivity of aquatic ecosystems. In this kit, students familiarize themselves with the Winkler method by examining the relationship between temperature and dissolved oxygen concentrations. Using local water samples to examine primary productivity at different water depths, students can then observe the beginning stages of eutrophication.

Order Info

Class size: 10 laboratory groups (40 students).

Specifications

Grade 9 to Undergraduate
For Use With (Application) Experiment demonstrates the methodology used for measuring dissolved oxygen and determining the primary productivity of a natural body of water
Includes Sodium iodide, sodium hydroxide, manganese sulfate, starch solution, sodium thiosulfate, nitrogen enrichment solution, phosphorous enrichment reagents, comprehensive instructions, background material and study questions
Time Range 2 Hr.
Content And Storage Room temperature
Class Size 40 Students
Product Type AP Biology Lab Kit 12: Dissolved Oxygen and Aquatic Primary Productivity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.