Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Advanced Placement Laboratory Kit 2: Principles of Enzyme Catalysis
SDPSCI_ED

Catalog No. s11282 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11282 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11282 Supplier Edvotek™ Supplier No. 282
Only null left
Add to Cart
Add to Cart

Includes

  • Catalase solution
  • 1.2% Hydrogen peroxide (stabilized)
  • Phosphate buffer, pH 7.2 (20x concentrate)
  • Assay reagent (potassium iodide concentrate)
  • Acidification solution, 0.1M HCl
  • Color enhancer (concentrate)
  • Color developer (concentrate)
  • Plastic tube, 15mL
  • Comprehensive instructions, background material, and study questions

Understand enzyme catalysis and protein structure-function relationships.

Methodology uses a colorimetric assay to measure the concentration of hydrogen peroxide (and therefore catalase activity) based on the oxidation of iodide by peroxide. This method differs from the AP Biology lab manual; it uses a safer system that eliminates the need for sulfuric acid.

  • Perform an enzyme assay
  • Determine the rate of a biochemical reaction

Specifications

Grade 9 to Undergraduate
For Use With (Application) Understand enzyme catalysis and protein structure function relationships
Includes Catalase solution, 1.2% Hydrogen Peroxide, 20X pH 7.2 Phosphate Buffer, Potassium Iodide concentrate assay reagent, 0.1M HCl acidification solution, color enhancer, color developer, 15mL Plastic tube, comprehensive instructions, background material and study questions
Time Range 30 to 45 min.
Content And Storage Refrigerated
Class Size 40 Students
Product Type AP Biology Lab Kit 2: Principles of Enzyme Catalysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.