Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Advanced Placement Laboratory Kit 4: Plant Pigment Chromatography and Photosynthesis
SDPSCI_ED

Catalog No. s11284 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11284 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11284 Supplier Edvotek™ Supplier No. 284
Only null left
Add to Cart
Add to Cart

Includes

instructions, chloroplast extraction; dilution buffers, reducing reagent, DCIP, standard pigments, chromatography solvent, Sarkosyl, thin layer cellulose chromatography plate, microliter capillary pipets, tubes and transfer pipets.

Requires

  • Visible wavelength spectrophotometer (see S42669ND)
  • Pencils
  • Clinical benchtop centrifuge
  • Metric rulers
  • Test tube racks
  • Flood light or small portable lamps (60w is sufficient)
  • Pipet pumps or bulbs
  • Distilled water
  • Test tubes, 10mL (13 x 100mm)
  • Beakers, 100mL
  • Pipets, 1 and 5mL
  • Ice
  • Cheesecloth
  • 95-100% isopropanol† (2-propanol)
  • Aluminum foil
  • Fresh spinach

Experiment demonstrates light-dependent reactions of photosynthesis by colorimetric assay, and helps students to gain an understanding of the biochemical processes by which they work.

Reformulated solvent systems are less flammable and less toxic than commonly used ether and acetone systems.

  • Gain an understanding of the theory and applications of chromatography
  • Use thin layer chromatography to separate plant pigments carotene, xanthophyll, and chlorophyll
  • Perform an assay of the light reactions of photosynthesis using intact chloroplasts and the dye DCIP

Order Info

Class size: 10 laboratory groups (40 students).

† Rubbing alcohol will not work, as it is only 70% isopropanol.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Experiment demonstrates light-dependent reactions of photosynthesis by colorimetric assay and helps students to gain an understanding of the biochemical processes by which they work
Includes Chloroplast extraction buffer, reducing reagent, chloroplast dilution buffer, dcip, standard pigments, chromatography solvent, sarkosyl, thin layer cellulose chromatography plate, 40 x 5μl capillary pipets, 4 plastic transfer pipets, 24
Time Range 1.75 hr.
Content And Storage Refrigerated
Class Size 40 Students
Product Type AP Biology Lab Kit 4: Plant Pigment Chromatography and Photosynthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.