Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Affinity Chromatography of Glucose-Binding Proteins
SDPSCI_ED

Catalog No. S05653 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05653 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05653 Supplier Edvotek™ Supplier No. 277
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructions
  • Affinity gel
  • Jack bean meal
  • NaCl, Dextrose
  • Con A Control
  • Horseradish peroxidase
  • ABTS Substrate
  • Standard dilution buffer
  • Membrane
  • Columns (syringe) and column tips
  • Cheesecloth for column
  • 50 ml conical tubes
  • Transfer pipets
  • Petri dishes

Isolate a carbohydrate-binding lectin protein from an extract of jack bean meal.

Affinity chromatography is applied to the purification of biologically active molecules. A ligand is coupled to an insoluble support. The specific protein binds to the ligand, which results in the separation of a specific protein from a mixture.

  • Students prepare a seed extract, fractionate the extract by affinity chromatography and elute the bound glucose-binding protein
  • As a second experiment, the presence of biological activity is determined by an immunoblot enzyme assay
  • Requires two hours

All You Need

  • shaking platform or vortex
  • rotating or rocking platform
  • clinical centrifuge
  • ring stands with clamps for columns
  • microteset tubes or small glass test tubes for collecting fractions
  • test tubes (15 ml) to collect elutant
  • 50 ml
  • 100 ml beakers or flasks
  • beaker or flask
  • graduated cylinders: 10 ml
  • 100 ml
  • and 250 ml
  • 10 ml pipets
  • forceps
  • distilled water
  • filter paper.

Order Info

For 10 groups.

Recommended Storage

Some components require refrigerator storage

Specifications

Grade 9 to Undergraduate
For Use With (Application) For Purification of Biologically Active Molecules
Includes Instructions, affinity gel, jack bean meal, various solutions and buffers, petri plates, columns with tips, conical tubes, membranes, and transfer pipets
Class Size 10 Lab Groups
Product Type Affinity Chromatography Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.