Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Amplification of DNA by PCR
SDPSCI_ED

Catalog No. S11330 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11330 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11330 Supplier Edvotek™ Supplier No. 330
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, PCR EdvoBeads™, LyphoPrimer™ Mix, DNA Standard Marker, Ultra-pure Water, LyphoTemplate™, TE Buffer, UltraSpec-Agarose™, 50X Electrophoresis Buffer, 10X Gel Loading Solution, FlashBlue™ Stain, InstaStain™ Ethidium Bromide, Microcentrifuge & PCR Tubes, Wax Beads

Easy PCR experiment, students will make billions of copies of a small amount of DNA.

In this easy PCR experiment, students will make billions of copies of a small amount of DNA in just 90 minutes! They will just need to mix template DNA & primers with PCR beads that contain all of the other components required to carry out a PCR reaction. Students will see the increasing amounts of DNA for themselves, taking samples every few cycles and analyzing them on a DNA gel.

Specifications

Grade 9 to Undergraduate
For Use With (Application) For billions of copies of a small amount of dna
Includes Instructions, pcr edvobeads™, dna template and primers, dna size ladder, ultrapure water, wax beads, gel loading dye, agarose, electrophoresis buffer, instastain™ ethidium bromide and flashblue™ stain
Requires 5 to 50μL adjustable micropipets, tips, thermal cycler, electrophoresis apparatus, power supply, microwave or hot plate, UV transilluminator
Time Range 3 Hr.
Class Size 10 Lab Groups
Product Type PCR Amplification of DNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.