Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Antigen-Antibody Interaction: The Ouchterlony Procedure Kit
SDPSCI_ED

Catalog No. S90246ND Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90246ND 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90246ND Supplier Edvotek™ Supplier No. 270
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Separate samples containing animal anti-serum, animal serum, albumin antigen and IgG antigen
  • Package of powdered buffer and package of UltraSpec-Agarose™
  • Tube of practice loading solution
  • 40 transfer pipets
  • 40 Petri dishes
  • 10 well cutters
  • Template for cutting wells
  • 40 microtest tubes

Requires

  • Micropipet and tips
  • Plastic container or glass baking dish
  • Plastic wrap
  • Distilled water
  • Pipets, 5 or 10mL
  • Marking pen
  • Measuring spatula or toothpicks
  • Hotplate, Bunsen burner or microwave
  • Paper towels
  • Water bath (55°C [131°F])

Introduce students to the principles of antigen-antibody interactions using the Ouchterlony procedure.

  • Demonstrates a fundamental reaction of immunology — the binding interaction of an antibody to an antigen and its resulting precipitate, which makes it possible to assay

No components have been prepared from human sources.

Order Info

Class size: 10 lab groups.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Introduce students to the principles of antigen antibody interactions using the ouchterlony procedure
Includes Instructions, animal serum antigens and antibodies, practice gel loading solution, agarose, powdered buffer, transfer pipets, petri plates, well cutters, microtest tubes
Time Range 35 min.
Content And Storage Refrigerated; Store at: 4°C; 39°F
Class Size 10 Lab Groups
Product Type Ouchterlony Procedure Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.