Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP™ LAB 1: Artificial Selection
SDPSCI_ED

Catalog No. S06879 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06879 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S06879 Supplier Edvotek™ Supplier No. AP01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Quick Plant seeds, nylon mason twine, potting mix, Miracle-Gro™ fertilizer, vermiculite, bees, plastic magnifier, wooden toothpicks and instructions

Requires

Growing System: reused plastic soda or water bottles (500mL), lighting, light box system, digital cameras, lab notebook, water, tape and razor

Students learn how artificial selection works.

Students will perform artificial selection on a population of Quick Plant™ and identify traits that vary in the population. Then they will perform artificial selection by cross-pollinating only selected plants and observe the trait differences between the two populations to learn how selection works.

Order Info

  • Class size: 10 laboratory groups
  • Takes 30-minute lab periods over course of 5 to 7 weeks to complete the process

Specifications

Grade 9 to 12
For Use With (Application) For pollination
Includes Instructions, quick plant™ seeds, nylon mason twine, potting mix, miracle-gro fertilizer, vermiculite, bees, plastic magnifier and wooden toothpicks
Requires Growing system: reused plastic soda or water bottles (500mL), lighting, light box system, digital cameras, lab notebook, water, tape, razor
Time Range 30 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Investigation 1: Artificial Selection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.