Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP LAB 13: Enzyme Activity
SDPSCI_ED

Catalog No. S06891 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06891 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06891 Supplier Edvotek™ Supplier No. AP13
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Hydrogen peroxide solution, guaiacol solution, phosphate buffer pH 3, phosphate buffer pH 7, phosphate buffer pH 10 and phosphate buffer pH 14

Requires

Turnip root, distilled or dionized water, pipet pumps or bulbs, Erlenmeyer flask (500mL), spectrophotometer, water baths, filter paper and funnel, test tube racks, test tubes (13 x 150mm), thermometer, cheesecloth, Parafilm™ film, hot plate, timer or clock with second hand, lab permanent markers, ice, razor, goggles and blender

Learn about enzyme catalysis and the nature of enzyme action.

This safe, easy experiment allows your students to learn about enzyme catalysis, the nature of enzyme action and protein structure-function relationships.

  • Students perform an enzyme assay and determine the rate of the enzymatic reaction Experiment uses a safer system that eliminates the need for sulfuric acid and potassium permanganate

Order Info

  • Class size: 10 lab groups
  • Requires 30 to 45 minutes

Specifications

Grade 9 to 12
For Use With (Application) To perform an enzyme assay and determine the rate of the enzymatic reaction
Includes Instructions, hydrogen peroxide solution, guaiacol solution, phosphate buffer ph 3, phosphate buffer ph 7, phosphate buffer ph 10, and phosphate buffer ph 14
Time Range 30 to 45 min.
Content And Storage Refrigerated
Class Size 10 Lab Groups
Product Type Enzyme Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.