Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP LAB 4: Diffusion and Osmosis
SDPSCI_ED

Catalog No. s06882 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06882 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06882 Supplier Edvotek™ Supplier No. AP04
Only null left
Add to Cart
Add to Cart

Includes

Agar powder, phenolphthalein solution, sodium hydroxide (NaOH) pellets, powdered sucrose, NaCl, powdered glucose, ovalbumin, dialysis tubing, large transfer pipets, microscope slides and cover slips

Requires

Beaker, ruler, razor, plastic spoon, paper towel, timer, scales, graph paper, distilled or deionized water, elodea tip or Moss and microscope

Students explore a variety of diffusion and osmosis principles in this lab.

In this experiment, students use artificial cells to study the relationship of surface area and volume. Then they will create models of living cells to explore osmosis and diffusion, and observe osmosis in living cells.

Order Info

  • Class size: 10 lab groups
  • Takes 60 to 90 minutes to complete

Specifications

Grade 9 to 12
For Use With (Application) Students study the relationship of surface area and volume
Includes Agar powder, phenolphthalein solution, sodium hydroxide (naoh) pellets, powdered sucrose, nacl, powdered glucose, ovalbumin, dialysis tubing, large transfer pipets, microscope slides and cover slips
Time Range 60 to 90 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Diffusion and Osmosis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.