Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP LAB 5: Photosynthesis
SDPSCI_ED

Catalog No. S06883 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06883 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06883 Supplier Edvotek™ Supplier No. AP05
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, Sodium Biocarbonate (baking soda), liquid soap, plastic syringe (20 cc), transfer pipets, plastic cups

Requires

Leaves, timer, light source, hand-held hole punch, beakers

Key reagents are included to demonstrate photosynthesis.

In this experiment, students will learn how to measure the rate of photosynthesis indirectly by studying the floating leaf disk assay, and test different variables that might affect the photosynthesis process.

Order Info

  • Class size: 10 lab groups
  • Takes one hour, 45 minutes over two lab periods to complete

Specifications

Grade 9 to 12
For Use With (Application) Students will learn how to measure the rate of photosynthesis indirectly by studying the floating leaf disk assay and test different variables that might affect the photosynthesis process
Includes Sodium biocarbonate (baking soda), liquid soap, plastic syringe (20 cc), transfer pipets, plastic cups, metric rulers, instructions, chloroplast extraction and dilution buffers, reducing agent, dcip, standard pigments
Requires Leaves (spinach, ivy, pokeweed), timer, light source (60w light bulb recommended), hand-held hole punch and beakers
Time Range 60 to 90 hr.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type AP Lab 5: Photosynthesis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.