Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP LAB 6: Cellular Respiration
SDPSCI_ED

Catalog No. s06884 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06884 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06884 Supplier Edvotek™ Supplier No. AP06/285
Only null left
Add to Cart
Add to Cart

Includes

instructions, 1 ml pipet, glass beads, peas, potassium hydroxide solution, cork stoppers, absorbent cotton, nonabsorbent cotton, plastic vials, parafilm

Requires

10 thermometers, 10 trays (at least 14 in. [35.6cm] long), silicon glue, ice, cork borer, tape and timers

Students use small respirometers to determine O2 consumption.

In this experiment, students learn how to apply the gas laws to the function of the microrespirometer. Students will observe cell respiration of germinating seeds and describe the effects of temperature on the rate of cell respiration.

Order Info

  • Class size: 10 lab groups
  • Requires two hours over two lab periods

Specifications

Grade 9 to 12
For Use With (Application) Students learn how to apply the gas laws to the function of the microrespirometer
Includes Instructions, 1ml pipet, glass beads, peas, potassium hydroxide solution, cork stoppers, absorbent cotton, nonabsorbent cotton, plastic vials, parafilm
Time Range 2 Hr.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Cellular Respiration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.