Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP LAB 9: Biotechnology — Restriction Enzyme Analysis of DNA
SDPSCI_ED

Catalog No. S06887 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06887 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06887 Supplier Edvotek™ Supplier No. AP09
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, DNA samples (packaged as either Pre-aliquoted QuickStrip™ connected tubes or as individual 1.5 ml (or 0.5 ml) microcentrifuge tubes), UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets.

Requires

Horizontal gel electrophoresis apparatus; DC power supply; automatic micropipets with tips; balance; microwave; hotplate or burner; pipet pump; 250mL flasks or beakers; hot gloves; safety goggles; disposable laboratory gloves; small plastic trays or large weigh boats (for gel destaining); DNA visualization system (white light) and distilled or deionized water

For analysis of Eco RI cleavage patterns of lambda DNA

  • This experiment introduces the use of restriction enzymes as a tool to digest DNA at specific nucleotide sequences
  • Bateriophage lambda DNA has a linear structure and six Eco RI recognition sites
  • Separation by agarose gel electrophoresis of an Eco RI digest of lambda DNA will yield six bands (five distinct bands, two are very close in size) corresponding to the DNA fragments

Order Info

  • For six gels
  • Takes one hour to complete

Specifications

Grade 9 to 12
For Use With (Application) Restriction enzyme analysis of dna
Includes Instructions, dna samples, ultraspec agarose™ powder, concentrated electrophoresis buffer, flashblue™ dna stain, instastain™ blue cards, practice gel loading solution, 1ml pipet, and microtipped transfer pipets
Quantity 8 Gels
Time Range 60 min.
Content And Storage Refrigerated
Class Size 10 Lab Groups
Product Type Biotechnology Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.