Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP™ Labs Special Package: 13 AP Labs
SDPSCI_ED

Catalog No. S06892 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06892 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06892 Supplier Edvotek™ Supplier No. APPKG
Add to Cart
Add to Cart

Help your high school students prepare for higher education as they learn the core concepts of this innovative and exciting introductory level college course.

Contains:

  • Lab 1: Artificial Selection
  • Lab 2: Mathematical Modeling: Hardy-Weinberg
  • Lab 3: Comparing DNA Sequences to Understand Evolutionary Relationships with BLAST
  • Lab 4: Diffusion and Osmosis
  • Lab 5: Photosynthesis
  • Lab 6: Cellular Respiration
  • Lab 7: Cell Division: Mitosis and Meiosis
  • Lab 8: Biotechnology: Bacterial Transformation
  • Lab 9: Biotechnology: Restriction Enzyme Analysis of DNA
  • Lab 10: Energy Dynamics
  • Lab 11: Transpiration
  • Lab 12: Fruit Fly Behavior
  • Lab 13: Enzyme Activity

Specifications

Grade 9 to 12
Includes Lab 1: Artificial Selection (Mfr. No. AP01); Lab 2: Mathematical Modeling: Hardy-Weinberg (Mfr. No. AP02); Lab 3: Comparing DNA Sequences to Understand Evolutionary Relationships with BLAST - Free Download (Mfr. No. AP03); Lab 4: Diffusion and Osmosis (Mfr. No. AP04); Lab 5: Photosynthesis (Mfr. No. AP05); Lab 6: Cellular Respiration (Mfr. No. AP06); Lab 7: Cell Division: Mitosis and Meiosis (Mfr. No. AP07); Lab 8: Biotechnology: Bacterial Transformation (Mfr. No. AP08); Lab 9: Biotechnology: Restriction Enzyme Analysis of DNA (Mfr. No. AP09); Lab 10: Energy Dynamics - Free Download (Mfr. No. AP10); Lab 11: Transpiration (Mfr. No. AP11); Lab 12: Fruit Fly Behavior (Mfr. No. AP12) Lab 13: Enzyme Activity (Mfr. No. AP13)
Class Size 10 Lab Groups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.