Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ BactoBeads™ Microorganism Cultures
SDPSCI_ED

Catalog No. S07093 Shop All EDVOTEK Products
Change view
Click to view available options
Species:
Bacillus subtilis
Micrococcus luteus
Serratia marcescens
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
S07093 Bacillus subtilis
S07091 Serratia marcescens
S07092 Micrococcus luteus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S07093 Supplier Edvotek™ Supplier No. 743
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use a BactoBead on an agar plate to streak for isolated colonies.

  • Used to streak for isolated colonies

Specifications

Biosafety Level 1
Product Type Tubed Media
Sterility Non-sterile
For Use With (Application) Classroom, research use
Species Bacillus subtilis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.