Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Blood-Based Cancer Diagnostics
SDPSCI_ED

Catalog No. S94672 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94672 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94672 Supplier Edvotek™ Supplier No. 141
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, microtiter plates, cancer cell markers, normal cell markers, transfer pipets, buffer

Requires

Distilled water

Use specific cell surface protein markers of cancer cells for their identification.

Cancer cells differ from normal cells by the combinations of proteins that are present on their surfaces. Antibodies against these proteins will specifically bind to cancer cells and not to normal cells. This allows early detection of cancer and potentially a way of delivering cancer therapies. In this simulation experiment the reaction of cancer cell markers and their corresponding antigens are demonstrated.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Specific cell surface protein markers of cancer cells for their identification
Includes Instructions, microtiter plates, cancer cell markers, normal cell markers, transfer pipets, buffer
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Blood-based Cancer Diagnostics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.