Learn More
Description
Includes
Enough supplies for 10 student groups
Specifications
Specifications
| Grade | 9 to Undergraduate |
| For Use With (Application) | To identify blood and perform rapid blood typing |
| Includes | Instructions, control abo simulated blood samples, simulated crime scene and suspect blood samples, anti-a and anti-b serums, blood detection stock solutions, transfer pipets, microtiter plates, tubes, filter paper, cotton swab |
| Requires | 95 to 100% ethanol, marking pen, distilled water |
| Content And Storage | Refrigerated |
| Class Size | 10 Lab Groups |
| Product Type | Forensics Blood Typing Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

