Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Blood Typing
SDPSCI_ED

Catalog No. S94671 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94671 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94671 Supplier Edvotek™ Supplier No. 140
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, control ABO simulated blood samples, unknown simulated blood samples, transfer pipets, microtiter plate

Learn about determining blood types.

  • Blood typing is essential for blood transfusion and other medical conditions.
  • Overview of blood typing is discussed
  • Hands-on determination of simple simulation reactions for blood ABO typing
  • Known and control samples provided for students to perform agglutination reactions to assign the various blood types
  • Class size: 10 groups
  • No blood or blood products are used in this experiment.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Learn about determining blood types
Includes Instructions, control abo rh simulated blood samples, unknown simulated blood samples, transfer pipettes, microtiter plate
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Blood Typing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.