Learn More
Edvotek™ Blue/White Cloning of DNA Fragment and Assay of β-galactosidase
SDPSCI_ED
Catalog No. S11300ND Shop All EDVOTEK Products
Change view
Click to view available options
Format:
Kit
Catalog No. Format
S11300ND Kit
1 options
Catalog No. S11300ND Supplier Edvotek™ Supplier No. 300
Add to Cart
Add to Cart
Includes
instructions, Linearized pUC plasmid & DNA fragment, T4 Ligase, BactoBeads™ for transformation, reconstitution buffer, XGal in solvent, IPTG, calcium chloride, antibiotic, ReadyPour™ Luria Broth Agar, Luria broth media for recovery, growth media, assay components, plastic supplies
Requires
Incubation oven, two waterbaths, shaking incubator or shaking waterbath, microwave or hot plate, automatic micropipet and tips, spectrophotometer, balance, centrifuge, microcentrifuge, glassware and cuvettes, distilled water and ice
To clone a DNA fragment in pUC-linker and select colonies that have DNA inserts based on color selection.
When DNA is subcloned in the pUC polylinker region, ß-galactosidase production is interrupted, resulting in the inability of cells to hydrolyze X-Gal. This results in the production of white colonies amongst a background of blue colonies. This experiment provides a DNA fragment together with a linear plasmid and T4 DNA Ligase. Following the ligation to synthesize the recombinant plasmid, competent E. coli cells are transformed and the number of recombinant antibiotic resistant white and blue colonies are counted. ß-galactosidase activity is assayed from blue and white bacterial cells. This experiment can be broken down into three modules: ligation, transformation, and assay of ß-galactosidase.
Specifications
Format Kit
Includes Instructions, linearized pUC plasmid and DNA fragment, T4 Ligase, bacterial LyphoCells™ for Transformation, Reconstitution Buffer, X-Gal in solvent, IPTG, Calcium Chloride, antibiotic, ReadyPour Luria Broth Agar, Luria broth media for re
Product Type Ligation, Transformation, and Assay of ß-galactosidase