Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Cleavage of Lambda DNA with EcoRI
SDPSCI_ED

Catalog No. S05644 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05644 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05644 Supplier Edvotek™ Supplier No. 212
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructions
  • Lambda phage DNA
  • Concentrated Restriction Enzyme Reaction Buffer
  • EDVOTEK™ Enzyme Grade Water
  • Eco RI Dryzymes™ endonuclease
  • DNA Standard Marker, Reconstitution Buffers
  • UltraSpec-Agarose™
  • 10X Gel Loading Solution
  • 50X Concentrated Electrophoresis Buffer
  • Practice Gel Loading Solution
  • FlashBlue™ DNA Stain
  • InstaStain™ Blue Cards
  • Disposable Pipets
  • Microtest Tubes
  • Semi-log Graph Paper Template

Requires

  • Electrophoresis apparatus
  • Power supply
  • Automatic pipet with tips
  • Water bath
  • Balance
  • Microwave or hotplate
  • Visualization (white light)
  • Lab glassware
  • Pipet pumps or bulbs
  • Metric rulers
  • Floating racks
  • Distilled or deionized water
  • Ice

The DNA from bacteriophage lambda is a well-characterized linear molecule containing six recognition sites for EcoRI (five distinct sites; two very close in size).

  • Students digest lambda DNA with EcoRI endonuclease
  • Digestion products are analyzed by agarose gel electrophoresis
  • Complete in 90 minutes

Order Info

Contains materials for 10 restriction digestions and five gels.

Recommended Storage

Some components require freezer storage

Specifications

Grade 9 to Undergraduate
For Use With (Application) Digestion products are analyzed by agarose gel electrophoresis
Includes Instructions, lambda dna, dryzymes™, reconstitution buffer, restriction enzyme reaction buffer, enzyme grade water, standard dna fragments, various solutions and buffers, agarose powder, flashblue™ stain
Quantity 10 Restriction Digestions and 5 Gels
Time Range 90 min.
Content And Storage Refrigerated; Store at: -20°C; -4°F
Class Size 5 Lab Groups
Product Type Cleavage of Lambda DNA with Eco Ri Endonuclease Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.