Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Clinical Diagnostic Immunoblot
SDPSCI_ED

Catalog No. S96347 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96347 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96347 Supplier Edvotek™ Supplier No. 276
Add to Cart
Add to Cart

Includes

  • Antigen
  • Antibody to antigen
  • IgG-horseradish peroxidase conjugate
  • Peroxidase substrate
  • Hydrogen peroxide
  • Substrate dilution buffer
  • Tris buffered saline (pH 7.5)
  • Instant nonfat dry milk
  • Nylon membranes
  • Petri dishes (60mm diameter)
  • Complete instructions, background information and study questions

Requires

  • Forceps
  • Graph paper
  • Rulers
  • Scissors
  • Shaking platform (optional)
  • Distilled water

Demonstrates a sensitive immunological assay used in research and clinical laboratories to quantify protein antigens.

Clinical diagnostics kits employ the principles of the immunoblot (dot blot), a technique used to determine the presence of an antigen.

  • In this experiment, students will prepare a dot blot membrane to determine the presence of an antigen
  • Antigens are absorbed into a membrane that is then treated sequentially with an antigen-specific antibody solution and a secondary antibody conjugated to an enzyme
  • The enzyme-substrate reaction generates a color product that precipitates on the membrane, indicating a positive reaction

Order Info

Class size: 10 groups.

Specifications

Grade 9 to Undergraduate
For Use With (Application) To determine the presence of an antigen
Includes Instructions, antigen, antibodies, various reagents and buffers, instant nonfat dry milk, hydrogen peroxide, nylon membranes and petri dishes
Time Range 1.5 to 2 hr.
Content And Storage Refrigerated; Store at: 4°C; 39°F
Class Size 10 Lab Groups
Product Type Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.