Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Demonstration DNA Electrophoresis LabStation™
SDPSCI_ED

Catalog No. s68644 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
4 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S68644 4 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68644 Supplier Edvotek™ Supplier No. 5061
Only null left
Add to Cart
Add to Cart

Includes

(1) M12 Complete™ Electrophoresis Package (1) DuoSource™ 75 Power Supply (75 V for 1 or 2 units) (1) Fixed Volume MiniPipet (35 μl) (1) Yellow Micropipet Tips(1 - 200 μl /2 Racks of 96) (1) EDVO-Kit #S-51 Whose DNA Was Left Behind?

Basic electrophoresis unit suitable for instructor-led presentations.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Electrophoresis
Includes M12 Complete™ Electrophoresis Package (Mfr. No. 502/504 ); DuoSource™ 75 Power Supply (Mfr. No.507); Fixed Volume MiniPipette 35 μL (Mfr. No.587-2 ); Yellow Micropipet Tips, 1 - 200 μL /2 Racks of 96 (Mfr. No. 636 ); Whose DNA Was Left Behind? Kit (Mfr. S-51 )
Class Size 4 Students
Product Type LabStation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.