Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Detecting the Silent Killer: Clinical Diagnosis of Diabetes
SDPSCI_ED

Catalog No. S21280 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S21280 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S21280 Supplier Edvotek™ Supplier No. 280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diagnose diabetes in three patients using urine glucose test and Enzyme-linked Immunosorbent Assay (ELISA).

  • For 10 lab groups
  • Complete in 90 minutes
  • Includes: Instructions, samples, antigens and antibodies, various solutions and reagents, pipettes and microtest tubes
  • Materials needed: 37°C incubation oven, waterbath, automatic micropipets with tips, laboratory glassware, distilled or deionized water
  • Storage: Some components require refrigerator storage

Specifications

Grade 9 to 12
For Use With (Application) Diagnose diabetes in patients using urine glucose test and Enzyme-linked Immunosorbent Assay (ELISA)
Class Size 10 Lab Groups
Product Type Detecting the Silent Killer: Clinical Diagnosis of Diabetes Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.