Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Detection of Genetically Modified Organisms Kit
SDPSCI_ED

Catalog No. s21121 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S21121 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S21121 Supplier Edvotek™ Supplier No. 121
Only null left
Add to Cart
Add to Cart

Use of agarose gel electrophoresis to explore molecular methods used by scientists to identify genetically modified organisms.

  • No thermal cycler is required
  • Students are encouraged to explore controversy surrounding use of genetically modified organisms
  • For 8 gels
  • Completes in 60 minutes
  • Includes: Instructions, Ready-to-Load™ QuickStrip™ DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain™ Blue and FlashBlue™ stain, calibrated pipette, and microtipped transfer pipettes
  • Materials needed: Electrophoresis apparatus, power supply, automatic micropipet and tips, balance, microwave or hot plate, white light visualization system
  • Storage: Stable at room temperature. Recommended to store Ready-to-Load QuickStrip™ samples in refrigerator

Specifications

Grade 9 to Undergraduate
For Use With (Application) Explore molecular methods used by scientists to identify genetically modified organisms
Class Size 8 Lab Groups
Product Type Detection of Genetically Modified Organisms Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.