Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Detection of Mad Cow Disease
SDPSCI_ED

Catalog No. S95607 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S95607 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95607 Supplier Edvotek™ Supplier No. 117
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets

Requires

Horizontal gel electrophoresis apparatus; DC power supply; Automatic micropipets with tips; Balance; Microwave, hotplate or burner; Pipet pump; 250mL flasks or beakers; Hot gloves; Safety goggles and disposable laboratory gloves; Small plastic trays or large weigh boats for gel destaining; DNA visualization system (white light); Distilled or deionized water

Educate students about mad cow disease.

Bovine spongiform encephalopathy (BSE), better known as mad cow disease, is a fatal neurodegenerative condition in cattle that can devastate the beef industry. One mode of BSE transmission throughout cattle populations is by the inclusion of infected cow or sheep parts in cattle feed, a practice that was banned in the United States in 1997.

  • Examine simulated PCR products from several feed mills
  • Determine any possible violations of the 1997 ban
  • Does not contain human DNA

Order Info

Contains materials for eight gels with InstaStain™ Methylene Blue.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Educate students about mad cow disease
Quantity 8 Gels
Content And Storage Refrigerated
Class Size 8 Lab Groups
Product Type Mad Cow Disease Detection Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.