Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Determination of Protein Molecular Weight (Polyacrylamide-based)
SDPSCI_ED

Catalog No. S05636 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05636 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05636 Supplier Edvotek™ Supplier No. 153
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions; Denatured LyphoProtein samples; Standard protein markers; Gel-loading solution; Buffer; Protein InstaStain™

Requires

Three polyacrylamide gels (12%); MV10 vertical gel electrophoresis apparatus; Power supply; Hotplate or burner; Visualization (white light); Automatic micropipet with fine tips; Microtest tube holder; Lab glassware; Methanol; Glacial acetic acid; Distilled or deionized water

Using prestained LyphoProteins™, subunit molecular weights are determined by analysis using denaturing SDS vertical polyacrylamide gel electrophoresis.

  • Prestained proteins with unknown molecular weights are assigned molecular weights based on the relative mobility of prestained standard protein markers

Experiment times:

  • Electrophoresis, one hour
  • Staining, 20 minutes
  • Destaining, two hours

Order Info

For six groups sharing three gels.

Recommended Storage

Some components require freezer storage

Specifications

Grade 9 to Undergraduate
For Use With (Application) Subunit molecular weights are determined by analysis using denaturing sds vertical polyacrylamide gel electrophoresis
Time Range 60 minutes; Staining time: 20 minutes
Content And Storage Refrigerated. Some components require freezer storage.
Class Size 6 Lab Groups
Product Type Protein Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.