Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Diversity of Fish Proteins
SDPSCI_ED

Catalog No. S05650 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05650 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05650 Supplier Edvotek™ Supplier No. 253
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, fish LyphoProtein™ samples, protein molecular weight standards, practice gel-loading solution, buffer and Protein InstaStain™

Determine the relationship between three fish species based on their protein profiles.

Study the diversity of fish with these prestained, lyophilized proteins.

  • Students extract and prestain total protein from perch, walleye and salmon using an indicator dye
  • After separating each fish protein sample by denaturing SDS-polyacrylamide gel electrophoresis, students find a characteristic banding pattern Pattern can be used to identify the specific species

Experiment times:

  • Electrophoresis, one hour
  • Staining, 20 minutes
  • Destaining, two hours

All You Need

  • 3 polyacrylamide gels (12%)
  • MV10 vertical gel electrophoresis apparatus
  • power supply
  • microcentrifuge
  • hot plate or burner
  • vortex
  • white light visualization system
  • automatic micropipet with fine tips
  • test tube holders
  • lab glassware
  • methanol
  • glacial acetic acid
  • distilled water.

Order Info

For six groups sharing three gels.

Recommended Storage

Some components require freezer storage

Specifications

Grade 9 to Undergraduate
For Use With (Application) Identify the specific species
Includes Instructions, fish lyphoprotein™ samples, protein molecular weight standards, practice gel loading solution, buffer, protein instastain™
Time Range 60 minutes; Staining time: 20 minutes
Content And Storage Refrigerated. Some components require freezer storage.
Class Size 6 Lab Groups
Product Type Protein Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.