Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA Bioinformatics Experiment
SDPSCI_ED

Catalog No. S67266 Shop All EDVOTEK Products
Change view
Click to view available options
DNA Bioinformatics Experiment
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67266 DNA Bioinformatics Experiment
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S67266 Supplier Edvotek™ Supplier No. 340

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Three sets of four autoradiograms
  • Complete instructions
  • Background information Study questions

Requires

  • Visualization (white light)
  • Computer access to the Internet

Access and learn from bioinformatics data.

In this experiment, students read autoradiographs containing DNA sequences which represent segments of important cellular genes. Using bioinformatics databases, students compare and extrapolate database information and identify the gene product

Order Info

Class size: 12 groups.

Specifications

Product Type DNA Bioinformatics Experiment
For Use With (Application) Read x-rays containing dna sequences
Includes Instructions, 3 sets of 4 autoradiograms, background information study questions
Requires White light visualization system, computer access to the internet
Time Range 45 min.
Class Size 12 Lab Groups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.