Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA Damage and Repair
SDPSCI_ED

Catalog No. S05694 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05694 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05694 Supplier Edvotek™ Supplier No. 957
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructions
  • Standard DNA fragments
  • Plasmid DNA
  • Gel-loading solution
  • Agarose
  • Electrophoresis buffer
  • 1mL pipet
  • Microtest tubes
  • 100mL graduated cylinder
  • InstaStain™ Ethidium Bromide

Requires

  • UV transilluminator
  • Electrophoresis apparatus
  • Power supply
  • Microwave or hotplate

Students can see firsthand the damage done by UV light.

According to the World Health Organization, between 2 and 3 million cases of skin cancer occur globally every year. Many of these cancers are caused by preventable damage to DNA by UV light during sunbathing.

  • Students expose plasmid DNA to shortwave UV light to simulate the effect of sunbathing
  • DNA is then analyzed by agarose gel electrophoresis to observe the damage
  • Complete in two hours

Order Info

For 10 groups.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Dna is analyzed by agarose gel electrophoresis to observe the damage
Includes Instructions, standard dna fragments, plasmid dna, gel loading solution, agarose, electrophoresis buffer, 1ml pipet, microtest tubes, 100ml graduated cylinder, instastain™ ethidium bromide™ stain
Time Range 2 Hr.
Content And Storage Some components require freezer storage
Class Size 10 Lab Groups
Product Type UV Damage Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.