Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA Fingerprinting Using PCR
SDPSCI_ED

Catalog No. S94592 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94592 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94592 Supplier Edvotek™ Supplier No. 371
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, PCR EdvoBeads™, Primer Mix Concentrate, EdvoQuick™ DNA Ladder, DNA Templates, TE Buffer, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, InstaStain™ Ethidum Bromide, FlashBlue™ DNA Stain, Microcentrifuge & PCR Tubes, & Wax Beads

Advanced experiments demonstrate PCR amplifications with easy-to-follow procedures.

  • Crime scene scenarios enable independent investigation of how Polymerase Chain Reaction (PCR) amplifications are used in DNA fingerprinting
  • Set of plasmids contains specifically designed DNA fragments
  • PCR amplification products obtained from the plasmids represent individual DNA profiles
  • Contains three advanced PCR experiments, each for 10 gels

Order Info

Class size: 20 to 30 students in groups of two or three.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Provides products that represent individual dna profiles
Includes Instructions; pcr edvobeads™; dna templates; primers; dna ladder; ultrapure water; wax beads; agarose; loading dye; electrophoresis buffer; instastain™ ethidium bromide and flashblue™ stain; TE buffer
Time Range 2 Hr.
Content And Storage Refrigerated; Store at: -20°C; -4°F
Class Size 25 Students
Product Type PCR-based DNA Fingerprinting Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.