Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA Fingerprinting Using Restriction Enzymes
SDPSCI_ED

Catalog No. s94610 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94610 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94610 Supplier Edvotek™ Supplier No. 225
Only null left
Add to Cart
Add to Cart

Includes

instructions, ''crime scene'' and ''suspect'' Ready-to-Load™ DNA samples, DNA Standard Marker, Dryzymes™ - Eco RI and Hind III, Enzyme Reaction Buffer, Reconstitution Buffer, Enzyme Grade Water, UltraSpec-Agarose™, 10X Gel Loading Solution, 50X Concentrated Electrophoresis Buffer, Practice Gel Loading Solution, InstaStain™ Blue Cards, FlashBlue™ DNA Stain, Disposable Pipets, & Microcentrifuge Tubes

Requires

Required: Horizontal gel electrophoresis apparatus; DC power supply; Automatic micropipets with tips; Water bathHotplate, Bunsen burner or microwave oven

Recommended: DNA visualization system (white light); Staining tray and net; 250mL flasks; Pipet pump; Hot gloves; Marking pens;Distilled or deionized water; Ice

Learn about restriction enzymes.

  • The objective of this simulated forensic analysis is to develop an understanding of DNA fingerprinting by use of restriction enzymes
  • Hypothetical crime scene samples are provided in addition to DNA samples, restriction enzymes and reaction buffer to digest suspect's DNA
  • Digestion products are separated by agarose gel electrophoresis
  • Students determine the guilty suspect through identification of the unique restriction enzyme fragmentation patterns
  • Does not contain human DNA
  • Some components require refrigeration, some require freezing

Order Info

Contains materials for six gels (six lanes each) with InstaStain™ Methylene Blue.

Specifications

Grade 9 to Undergraduate
For Use With (Application) For dna fingerprinting
Quantity 6 Gels
Requires Electrophoresis apparatus; power supply; automatic pipet with tips; waterbath; balance; microwave or hot plate; visualization (white light); misc. labware; pipet pumps or bulbs; metric rulers; floating racks; photodocumentation system (optional)
Time Range 1.45 hr.
Content And Storage Some components require refrigeration and some require freezing
Class Size 8 Lab Groups
Product Type DNA Fingerprinting Using Restriction Enzymes Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.