Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Do Onions, Strawberries and Bananas Have DNA?
SDPSCI_ED

Catalog No. S94595 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94595 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94595 Supplier Edvotek™ Supplier No. S75
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

DNA extraction buffer; DNA sample in capped test tube; Five transfer pipets; Pop beads for building DNA model; Glass rod; Four graduated test tubes; Five wood splints for DNA spooling; Salt packet; Complete instructions, background information and study questions

Requires

Fresh onion pieces; Ripened fresh banana; Fresh strawberry pieces; 70% clear isopropyl alcohol (rubbing alcohol); Distilled water

Students will participate in extracting DNA from fresh onion tissue.

  • Perform techniques used by scientists to explore the characteristics of DNA and see how DNA looks to the naked eye
  • Explain the function of DNA in cells
  • Build a simulated DNA model
  • Learn what DNA looks like in solution and after it is spooled
  • Isolate DNA from onion, banana or tomato cells and spool it from the solution

Order Info

Class size: five groups.

Specifications

Grade 6 to 12
For Use With (Application) For DNA extract
Includes DNA extraction Buffer; DNA sample in capped test tube; five transfer pipets; pop beads for building DNA model; Glass rod; four graduated test tubes; five wood splints for DNA spooling; salt packet; complete instructions; background information and study questions
Requires Fruit, vegetables and 95 to 100% isopropyl alcohol
Time Range 30 min.
Content And Storage Refrigerated
Class Size 10 Lab Groups
Product Type Do Onions, Strawberries and Bananas Have DNA? Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.